CAS RN 1784281-97-5
CAS RN 1784281-97-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Tocris Bioscience™ LDN 209929 dihydrochloride
Potent and selective haspin inhibitor
2
3