accessibility menu, dialog, popup

UserName

CH3CH3H3CSiCH3CH3Cl

CAS RN 18162-48-6

IUPAC Name: tert-butyl(chloro)dimethylsilane
Synonyms: t-butyldimethylchlorosilane
Molecular Weight (g/mol): 150.72
Molecular Formula: C6H15ClSi
InChi Key: BCNZYOJHNLTNEZ-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)Cl
Molecular Weight (g/mol) 
  • (5)
  • (6)
  • (4)
Quantity 
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (1)
  • (3)
  • (4)
Physical Form 
  • (4)
  • (2)
Boiling Point 
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (6)
  • (3)
  • (5)
  • (6)
  • (4)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (4)
  • (4)
  • (2)
  • (1)
  • (4)