accessibility menu, dialog, popup

UserName

ClCH2P

CAS RN 18480-23-4

IUPAC Name: triphenyl(prop-2-en-1-yl)phosphanium chloride
Synonyms: triphenyl(prop-2-en-1-yl)phosphanium chloride
Molecular Weight (g/mol): 338.81
Molecular Formula: C21H20ClP
InChi Key: FKMJROWWQOJRJX-UHFFFAOYSA-M
SMILES: [Cl-].C=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results