missing translation for 'onlineSavingsMsg'
Learn More

Allyltriphenylphosphonium chloride, 99%

Product Code. 11498626
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498626 25 g 25g
11408636 100 g 100g
11418636 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498626 Supplier Thermo Scientific Alfa Aesar Supplier No. A16012.14
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 18480-23-4 | C21H20ClP | 338.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18480-23-4
Molecular Formula C21H20ClP
Molecular Weight (g/mol) 338.81
MDL Number MFCD00031542
InChI Key FKMJROWWQOJRJX-UHFFFAOYSA-M
Synonym allyltriphenylphosphonium chloride, allyl triphenylphosphonium chloride, triphenyl prop-2-enyl phosphanium chloride, triphenyl prop-2-en-1-yl phosphanium chloride, acmc-1buz2, allyltriphenylphosphoniumchloride, triphenyl prop-2-enyl phosphonium chloride, phosphonium,triphenyl-2-propen-1-yl-, chloride 1:1
PubChem CID 2734120
SMILES [Cl-].C=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 230°C to 232°C
Linear Formula (CH2=CHCH2)(C6H5)3PCl
Quantity 25 g
Sensitivity Hygroscopic
IUPAC Name triphenyl(prop-2-enyl)phosphanium;chloride
Formula Weight 338.82
Percent Purity 99%
Chemical Name or Material Allyltriphenylphosphonium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.