accessibility menu, dialog, popup

UserName

ONOSH

CAS RN 1849-36-1

IUPAC Name: 4-nitrobenzene-1-thiol
Synonyms: 4-nitrobenzenethiol
Molecular Weight (g/mol): 155.17
Molecular Formula: C6H5NO2S
InChi Key: AXBVSRMHOPMXBA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(S)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (2)
Percent Purity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)