accessibility menu, dialog, popup

UserName

CH3H3CCH3OON(S)(R)O

CAS RN 185099-67-6

IUPAC Name: tert-butyl (1R,5S)-3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate
Synonyms: tert-butyl (1R,5S)-3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate
Molecular Weight (g/mol): 225.29
Molecular Formula: C12H19NO3
InChi Key: MENILFUADYEXNU-DTORHVGOSA-N
SMILES: CC(C)(C)OC(=O)N1[C@H]2CC[C@@H]1CC(=O)C2
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (2)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results