accessibility menu, dialog, popup

UserName

CAS RN 1871-22-3

HHClClH3CONHNNNNHNNNOCH3

CAS RN 1871-22-3

IUPAC Name: dihydrogen 2-[4'-(3,5-diphenyl-2,3-dihydro-1H-1,2,3,4-tetrazol-2-yl)-3,3'-dimethoxy-[1,1'-biphenyl]-4-yl]-3,5-diphenyl-2,3-dihydro-1H-1,2,3,4-tetrazole dichloride
Synonyms: dihydrogen 2-[4'-(3,5-diphenyl-1H-1,2,3,4-tetrazol-2-yl)-3,3'-dimethoxy-[1,1'-biphenyl]-4-yl]-3,5-diphenyl-1H-1,2,3,4-tetrazole dichloride
Molecular Weight (g/mol): 731.68
Molecular Formula: C40H36Cl2N8O2
InChi Key: RCEHREKDVGHYAM-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].COC1=CC(=CC=C1N1NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=C(N2NC(=NN2C2=CC=CC=C2)C2=CC=CC=C2)C(OC)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results