accessibility menu, dialog, popup

UserName

H3CONOOOH3C(E)

CAS RN 1891-10-7

IUPAC Name: 2,4-dimethoxy-1-[(1E)-2-nitroethenyl]benzene
Synonyms: 2,4-dimethoxy-1-[(1E)-2-nitroethenyl]benzene
Molecular Weight (g/mol): 209.2
Molecular Formula: C10H11NO4
InChi Key: NUMXHEUHHRTBQT-AATRIKPKSA-N
SMILES: COC1=CC=C(\C=C\[N+]([O-])=O)C(OC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results