accessibility menu, dialog, popup

UserName

H3COOH

CAS RN 18979-50-5

IUPAC Name: 4-propoxyphenol
Synonyms: phenol, 4-propoxy-
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: KIIIPQXXLVCCQP-UHFFFAOYSA-N
SMILES: CCCOC1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)