missing translation for 'onlineSavingsMsg'
Learn More

4-Propoxyphenol, 96%, Thermo Scientific™

Product Code. 10721681 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10721681 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10721681 Supplier Thermo Scientific Acros Supplier No. 213320500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18979-50-5
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00002335
InChI Key KIIIPQXXLVCCQP-UHFFFAOYSA-N
Synonym phenol, 4-propoxy, 4-n-propoxyphenol, p-propoxyphenol, phenol, p-propoxy, hydroquinone monopropyl ether, p-propoxy phenol, 4-propyloxyphenol, 4-propoxy-phenol, pubchem13204, para-propoxyphenol
PubChem CID 29352
IUPAC Name 4-propoxyphenol
SMILES CCCOC1=CC=C(C=C1)O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 55.0°C to 58.0°C
Flash Point >110°C
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula CH3CH2CH2OC6H4OH
Quantity 50 g
Formula Weight 152.19
Percent Purity 96%
Chemical Name or Material 4-Propoxyphenol, 96%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Propoxyphenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.