CAS RN 190850-49-8
CAS RN 190850-49-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Alpha-Ethyl-Alpha-(1-demethylethyl)-Verapamil Hydrochloride, TRC
CAS: 190850-49-8 Molecular Formula: C26H36N2O4.HCl Molecular Weight (g/mol): 477.04 Synonym: (2RS)-2-(3,4-Dimethoxyphenyl)-5-[[2-(3,4-dimethoxyphenyl)-ethyl](methyl)amino]-2-ethylpentanenitrile Hydrochloride,Verapamil Hydrochloride Imp. H (EP) as Hydrochloride IUPAC Name: 2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-ethylpentanenitrile;hydrochloride SMILES: Cl.CCC(CCCN(C)CCc1ccc(OC)c(OC)c1)(C#N)c2ccc(OC)c(OC)c2
2
(2RS)-2-(3,4-Dimethoxyphenyl)-5-[[2-(3,4-dimethoxyphenyl)ethyl](methyl)amino]-2-ethylpentanenitrile Hydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.