CAS RN 1914078-41-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
eCF506, MedChemExpress
MedChemExpress eCF506 is a highly potent and orally bioavailable inhibitor of the non-receptor tyrosine kinase Src with an IC50 of less than 0.5 nM.