missing translation for 'onlineSavingsMsg'
Learn More

eCF506, MedChemExpress

Product Code. 16497277
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497277 5 mg 5mg
16480867 50 mg 50mg
16455416 100 mg 100mg
16454568 10 mg 10mg
16433136 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16497277 Supplier Medchem Express Supplier No. HY112096

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress eCF506 is a highly potent and orally bioavailable inhibitor of the non-receptor tyrosine kinase Src with an IC50 of less than 0.5 nM.

eCF506 is a highly potent and orally bioavailable inhibitor of the non-receptor tyrosine kinase Src with an IC50 of less than 0.5 nM.

Specifications

Chemical Name or Material eCF506
CAS 1914078-41-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C26H38N8O3
Quantity 5 mg
Solubility Information DMSO : 62.5 mg/mL (122.40 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES NC1=C2C(C3=CC(OC)=C(NC(OC(C)(C)C)=O)C=C3)=NN(CCN4CCC(N(C)C)CC4)C2=NC=N1
Molecular Weight (g/mol) 510.63
Formula Weight 510.63
Percent Purity 99.17%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.