CAS RN 192185-72-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tipifarnib, TRC
CAS: 192185-72-1 Molecular Formula: C27 H22 Cl2 N4 O Molecular Weight (g/mol): 489.4 IUPAC Name: 6-[(R)-amino-(4-chlorophenyl)-(3-methylimidazol-4-yl)methyl]-4-(3-chlorophenyl)-1-methylquinolin-2-one SMILES: CN1C(=O)C=C(c2cccc(Cl)c2)c3cc(ccc13)[C@](N)(c4ccc(Cl)cc4)c5cncn5C
2
Tipifarnib, MedChemExpress
MedChemExpress Tipifarnib (IND 58359) binds to and inhibits farnesyltransferase (FTase) with an IC50 of 0.86 nM. Antineoplastic activity.