accessibility menu, dialog, popup

UserName

CAS RN 193620-69-8

H3COHNOOH3C

CAS RN 193620-69-8

IUPAC Name: 3-[bis(4-methoxyphenyl)methylidene]-2,3-dihydro-1H-indol-2-one
Synonyms: 3-[bis(4-methoxyphenyl)methylidene]-1H-indol-2-one
Molecular Weight (g/mol): 357.41
Molecular Formula: C23H19NO3
InChi Key: KUEYYIJXBRWZIB-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=C1C(=O)NC2=CC=CC=C12)C1=CC=C(OC)C=C1
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results