accessibility menu, dialog, popup

UserName

CH3H3CCH3NCO

CAS RN 1943-67-5

IUPAC Name: 1-tert-butyl-4-isocyanatobenzene
Synonyms: 1-tert-butyl-4-isocyanatobenzene
Molecular Weight (g/mol): 175.23
Molecular Formula: C11H13NO
InChi Key: WUWBDQJTQTVBSQ-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)N=C=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results