missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyl)phenyl isocyanate, 97%, Thermo Scientific™

Product Code. 10051144
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10051144 25 g Amber Glass Bottle 25g
10049551 1 g Amber Glass Bottle 1g
10079571 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10051144 Supplier Thermo Scientific Maybridge Supplier No. SB01802EB
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1943-67-5
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.231
MDL Number MFCD00040718
InChI Key WUWBDQJTQTVBSQ-UHFFFAOYSA-N
Synonym 4-tert-butylphenyl isocyanate, 4-tert-butylphenylisocyanate, benzene, 1-1,1-dimethylethyl-4-isocyanato, 4-tert-butylphenyl-isocyanate, 4-tert-butyl phenyl isocyanate, 1-tert-butyl-4-isocyanato-benzene, benzene,1-1,1-dimethylethyl-4-isocyanato, 4-t-butylphenylisocyanate, acmc-209ey5, 4-tertbutylphenyl isocyanate
PubChem CID 2735695
IUPAC Name 1-tert-butyl-4-isocyanatobenzene
SMILES CC(C)(C)C1=CC=C(C=C1)N=C=O

Specifications

CAS Min % 97.0
Density 0.986g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula (CH3)3CC6H4NCO
Quantity 25 g
Formula Weight 175.23
Percent Purity 97%
Chemical Name or Material 4-(Tert-butyl)phenyl isocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.