accessibility menu, dialog, popup

UserName

CAS RN 1946-82-3

H2N(S)NHOHOH3CO

CAS RN 1946-82-3

IUPAC Name: 6-amino-2-acetamidohexanoic acid
Synonyms: 6-amino-2-acetamidohexanoic acid
Molecular Weight (g/mol): 188.23
Molecular Formula: C8H16N2O3
InChi Key: VEYYWZRYIYDQJM-UHFFFAOYNA-N
SMILES: CC(=O)NC(CCCCN)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)