missing translation for 'onlineSavingsMsg'
Learn More

N Alpha-Acetyl-L-Lysine, TRC

Product Code. 30137220
Change view
Click to view available options
Quantity:
500 mg
1 g
25 g
Unit Size:
1g
25g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137220 25 g 25g
30132883 1 g 1g
30006559 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137220 Supplier Toronto Research Chemicals Supplier No. TRCA18554025G
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1946-82-3
Molecular Formula C8 H16 N2 O3
Molecular Weight (g/mol) 188.22
Synonym 6-Amino-L-2-acetamidohexanoic Acid, N2-Acetyl-L-lysine, N2-Acetyllysine, N-Acetyl-L-lysine, Nα-Acetyllysine
IUPAC Name (2S)-2-acetamido-6-aminohexanoic acid
SMILES CC(=O)N[C@@H](CCCCN)C(=O)O

Specifications

InChI Formula InChI=1S/C8H16N2O3/c1-6(11)10-7(8(12)13)4-2-3-5-9/h7H,2-5,9H2,1H3,(H,10,11)(H,12,13)/t7-/m0/s1
Quantity 25 g
Formula Weight 188.1161
Shipping Condition Room Temperature
Chemical Name or Material Nalpha-Acetyl-L-Lysine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.