missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nalpha-Acetyl-L-lysine, 99+%

Product Code. 10348080 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10348080 250 mg Glass bottle 250mg
10165000 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10348080 Supplier Thermo Scientific Chemicals Supplier No. 227632500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1946-82-3 | C8H16N2O3 | 188.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1946-82-3
Molecular Formula C8H16N2O3
Molecular Weight (g/mol) 188.23
MDL Number MFCD00008233
InChI Key VEYYWZRYIYDQJM-JLDDOWRYNA-N
PubChem CID 92907
ChEBI CHEBI:35704
IUPAC Name (2S)-2-acetamido-6-aminohexanoic acid
SMILES CC(=O)N[C@@H](CCCCN)C(O)=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 256°C to 258°C
Color White
Loss on Drying 0.5% max. (105°C, 4 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula H2N(CH2)4CH(NHCOCH3)CO2H
Quantity 250 mg
Specific Rotation + 5.6
Synonym ac-lys-oh, n-alpha-acetyl-l-lysine, n alpha-acetyllysine, nalpha-acetyl-l-lysine, n-acetyl-l-lysine, n2-acetyl-l-lysine, s-2-acetamido-6-aminohexanoic acid, n alpha-acetyl-l-lysine, n 2-acetyl-l-lysine, acetyl-l-lysine
Formula Weight 188.23
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Nα-Acetyl-L-lysine, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.