accessibility menu, dialog, popup

UserName

CH3OOOOH

CAS RN 19492-03-6

IUPAC Name: 8-hydroxy-7-methoxy-2H-chromen-2-one
Synonyms: 8-hydroxy-7-methoxychromen-2-one
Molecular Weight (g/mol): 192.17
Molecular Formula: C10H8O4
InChi Key: KIGCGZUAVODHMD-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=CC(=O)OC2=C1O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results