missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxy-7-methoxycoumarin, 97%, Thermo Scientific™

Product Code. 11383277
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11383277 100 mg 100mg
11373277 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383277 Supplier Thermo Scientific Alfa Aesar Supplier No. L13841.MC
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19492-03-6
Molecular Formula C10H8O4
Molecular Weight (g/mol) 192.17
MDL Number MFCD00075652
InChI Key KIGCGZUAVODHMD-UHFFFAOYSA-N
PubChem CID 146487
IUPAC Name 8-hydroxy-7-methoxychromen-2-one
SMILES COC1=C(C2=C(C=C1)C=CC(=O)O2)O

Specifications

Melting Point 175°C to 177°C
Odor Odorless
Quantity 100 mg
Beilstein 168393
Synonym 8-hydroxy-7-methoxycoumarin, daphnetin-7-methylether, 8-hydroxy-7-methoxy-2h-chromen-2-one, 8-hydroxy-7-methoxy-2h-1-benzopyran-2-one, 2h-1-benzopyran-2-one, 8-hydroxy-7-methoxy, 2h-1-benzopyran-2-one,8-hydroxy-7-methoxy, acmc-20alwe, daphnetin 7-methylether, daphnetin 7-methyl ether, daphnetin-7-methyl ether
Formula Weight 192.17
Percent Purity 97%
Chemical Name or Material 8-Hydroxy-7-methoxycoumarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.