CAS RN 1949837-12-0
CAS RN 1949837-12-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ ARV 771
Potent BET bromodomain PROTAC™; also degrades BRD-tagged chimeric antigen receptors (CAR) in T cells
2
ARV-771, MedChemExpress
MedChemExpress ARV-771 is a potent BET PROTAC based on E3 ligase von Hippel-Lindau with Kds of 34 nM, 4.7 nM, 8.3 nM, 7.6 nM, 9.6 nM, and 7.6 nM for BRD2(1), BRD2(2), BRD3(1), BRD3(2), BRD4(1), and BRD4(2), respectively.