accessibility menu, dialog, popup

UserName

CH3CH3H3COONFF

CAS RN 195447-25-7

IUPAC Name: tert-butyl 3,3-difluoropyrrolidine-1-carboxylate
Synonyms: tert-butyl 3,3-difluoropyrrolidine-1-carboxylate
Molecular Weight (g/mol): 207.22
Molecular Formula: C9H15F2NO2
InChi Key: ULFWNKDACVUZOD-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(F)(F)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results