accessibility menu, dialog, popup

UserName

CH3ONHSOONHClOCH3

CAS RN 196869-06-4

IUPAC Name: N-{4-[(5-chloro-2-methoxyphenyl)sulfamoyl]phenyl}acetamide
Synonyms: N-{4-[(5-chloro-2-methoxyphenyl)sulfamoyl]phenyl}acetamide
Molecular Weight (g/mol): 354.81
Molecular Formula: C15H15ClN2O4S
InChi Key: JVUHNUSVIURRRK-UHFFFAOYSA-N
SMILES: COC1=C(NS(=O)(=O)C2=CC=C(NC(C)=O)C=C2)C=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results