missing translation for 'onlineSavingsMsg'
Learn More

4'-(5-Chloro-2-methoxyphenylsulfamoyl)acetanilide, 97%, Thermo Scientific™

Product Code. 15456036
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15456036 250 mg 250mg
15446036 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15456036 Supplier Thermo Scientific Alfa Aesar Supplier No. H60820.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 196869-06-4
Molecular Formula C15H15ClN2O4S
Molecular Weight (g/mol) 354.805
MDL Number MFCD00026154
InChI Key JVUHNUSVIURRRK-UHFFFAOYSA-N
Synonym n-4-5-chloro-2-methoxyphenyl sulfamoyl phenyl acetamide, n-4-5-chloro-2-methoxyphenyl amino sulfonyl phenyl acetamide, cbmicro_023955, cambridge id 5536192, 4'-5-chloro-2-methoxyphenylsulfamoyl acetanilide, 4'-n-5-chloro-2-methoxyphenyl sulfamoyl acetanilide
PubChem CID 989525
IUPAC Name N-[4-[(5-chloro-2-methoxyphenyl)sulfamoyl]phenyl]acetamide
SMILES CC(=O)NC1=CC=C(C=C1)S(=O)(=O)NC2=C(C=CC(=C2)Cl)OC

Specifications

Quantity 250 mg
Formula Weight 354.81
Percent Purity 97%
Chemical Name or Material 4'-(5-Chloro-2-methoxyphenylsulfamoyl)acetanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.