accessibility menu, dialog, popup

UserName

CAS RN 1989-33-9

IUPAC Name: 9H-fluorene-9-carboxylic acid
Synonyms: 9-carboxyfluorene
Molecular Weight (g/mol): 210.23
Molecular Formula: C14H10O2
InChi Key: DNVJGJUGFFYUPT-UHFFFAOYSA-N
SMILES: OC(=O)C1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results