missing translation for 'onlineSavingsMsg'
Learn More

Fluorene-9-carboxylic acid, 97%

Product Code. 11430837
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430837 100 g 100g
11410837 5 g 5g
11420837 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430837 Supplier Thermo Scientific Alfa Aesar Supplier No. B23224.22
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1989-33-9 | C14H10O2 | 210.232 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1989-33-9
Molecular Formula C14H10O2
Molecular Weight (g/mol) 210.232
MDL Number MFCD00001136
InChI Key DNVJGJUGFFYUPT-UHFFFAOYSA-N
Synonym fluorene-9-carboxylic acid, 9-fluorenecarboxylic acid, 9-carboxyfluorene, diphenyleneacetic acid, 9-fluorencarboxylic acid, rarechem aq bd 0ba1, akos auf02021, 9-carboxy-9h-fluorene, enamine_003081, 9-fluorenylcarboxylic acid
PubChem CID 74809
IUPAC Name 9H-fluorene-9-carboxylic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)C(=O)O

Specifications

Melting Point 226°C to 232°C
Flash Point 230°C (446°F)
Quantity 100 g
Beilstein 1911728
Formula Weight 210.23
Percent Purity 97%
Chemical Name or Material Fluorene-9-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.