accessibility menu, dialog, popup

UserName

OHOONOOHOHO

CAS RN 2001-96-9

IUPAC Name: 2-(4-nitrophenoxy)oxane-3,4,5-triol
Synonyms: 2-(4-nitrophenoxy)oxane-3,4,5-triol
Molecular Weight (g/mol): 271.23
Molecular Formula: C11H13NO7
InChi Key: MLJYKRYCCUGBBV-UHFFFAOYNA-N
SMILES: OC1COC(OC2=CC=C(C=C2)[N+]([O-])=O)C(O)C1O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results