accessibility menu, dialog, popup

UserName

CAS RN 2021-28-5

IUPAC Name: ethyl 3-phenylpropanoate
Synonyms: ethyl B-phenylpropionate
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: JAGZUIGGHGTFHO-UHFFFAOYSA-N
SMILES: CCOC(=O)CCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results