missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-phenylpropionate, 98+%

Product Code. 11410004
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11410004 50 g 50g
11420004 250 g 250g
11400004 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410004 Supplier Thermo Scientific Alfa Aesar Supplier No. A13055.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2021-28-5 | C11H14O2 | 178.231 g/mol

Ethyl 3-phenylpropionate is used as an active pharmaceutical ingredient intermediate. It is also used in the synthesis of pyrimidine based inhibitors of cyclin-dependant kinases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-phenylpropionate is used as an active pharmaceutical ingredient intermediate. It is also used in the synthesis of pyrimidine based inhibitors of cyclin-dependant kinases.

Solubility
Miscible with oils and ethanol. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2021-28-5
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00009206
InChI Key JAGZUIGGHGTFHO-UHFFFAOYSA-N
Synonym ethyl 3-phenylpropionate, ethyl hydrocinnamate, ethyl dihydrocinnamate, benzenepropanoic acid, ethyl ester, 3-phenylpropionic acid ethyl ester, hydrocinnamic acid, ethyl ester, ethylhydrocinnamoate, ethyl benzenepropanoate, unii-58e9z9v64d, ethylphenyl propanoate
PubChem CID 16237
IUPAC Name ethyl 3-phenylpropanoate
SMILES CCOC(=O)CCC1=CC=CC=C1

Specifications

Density 1.016
Boiling Point 118°C (3 mmHg)
Flash Point 107°C (224°F)
Refractive Index 1.494
Quantity 50 g
Beilstein 1910432
Solubility Information Miscible with oils and ethanol. Immiscible with water.
Formula Weight 178.23
Percent Purity ≥98%
Chemical Name or Material Ethyl 3-phenylpropionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.