CAS RN 202842-98-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3,4-Dimethylpyrazole Phosphate, TRC
CAS: 202842-98-6 Molecular Formula: C5 H8 N2 . H3 O4 P Molecular Weight (g/mol): 194.13 Synonym: 3,4-Dimethyl-1H-pyrazole Phosphate,1H-Pyrazole, 3,4-dimethyl-, phosphate (1:1),3,4-Dimethyl-1H-pyrazolium dihydrogen phosphate,3,4-Dimethylpyrazole phosphate,DMPP,DMPP (nitrification inhibitor),Entec IUPAC Name: 3,4-dimethyl-1H-pyrazole;phosphoric acid SMILES: Cc1c[nH]nc1C.OP(=O)(O)O