missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylpyrazole Phosphate, TRC

Product Code. 30136446
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136446 5 g 5g
30135571 10 g 10g
17982375 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136446 Supplier Toronto Research Chemicals Supplier No. TRCD4789605G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202842-98-6
Molecular Formula C5 H8 N2 . H3 O4 P
Molecular Weight (g/mol) 194.13
Synonym 3,4-Dimethyl-1H-pyrazole Phosphate, 1H-Pyrazole, 3,4-dimethyl-, phosphate (1:1), 3,4-Dimethyl-1H-pyrazolium dihydrogen phosphate, 3,4-Dimethylpyrazole phosphate, DMPP, DMPP (nitrification inhibitor), Entec
IUPAC Name 3,4-dimethyl-1H-pyrazole;phosphoric acid
SMILES Cc1c[nH]nc1C.OP(=O)(O)O

Specifications

Analyte or Component Names 3,4-Dimethylpyrazole Phosphate
Shipping Condition Room Temperature
InChI Formula InChI=1S/C5H8N2.H3O4P/c1-4-3-6-7-5(4)2;1-5(2,3)4/h3H,1-2H3,(H,6,7);(H3,1,2,3,4)
Solution Type Neat
Industry Type Pharmaceutical
Quantity 5 g
Formula Weight 194.0456
Chemical Name or Material 3,4-Dimethylpyrazole Phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.