accessibility menu, dialog, popup

UserName

CAS RN 202865-73-4

FBrI

CAS RN 202865-73-4

IUPAC Name: 2-bromo-4-fluoro-1-iodobenzene
Synonyms: 2-bromo-4-fluoro-1-iodobenzene
Molecular Weight (g/mol): 300.9
Molecular Formula: C6H3BrFI
InChi Key: GKGPZAZTXZCUMU-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=C(I)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results