missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluoro-1-iodobenzene, 98+%

Product Code. 11355649
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11355649 1 g 1g
11344596 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11355649 Supplier Thermo Scientific Alfa Aesar Supplier No. L20144.03
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 202865-73-4 | C6H3BrFI | 300.897 g/mol

It is employed in the synthesis of tetrasubstituted alkenes by stereo- and regioselective stannyllithiation of diarylacetylenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of tetrasubstituted alkenes by stereo- and regioselective stannyllithiation of diarylacetylenes.

Solubility
Not miscible or difficult to mix in water.

Notes
Light sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202865-73-4
Molecular Formula C6H3BrFI
Molecular Weight (g/mol) 300.897
MDL Number MFCD00142868
InChI Key GKGPZAZTXZCUMU-UHFFFAOYSA-N
Synonym 2-bromo-4-fluoroiodobenzene, 2-bromo-4-fluoro-1-iodo-benzene, 2'-bromo-4-fluoro-1-iodobenzene, benzene, 2-bromo-4-fluoro-1-iodo, pubchem3234, acmc-1cll6, ksc495e7f, 2-bromo-4-fluoro-iodobenzene, 1-fluoro-3-bromo-4-iodobenzene
PubChem CID 2773372
IUPAC Name 2-bromo-4-fluoro-1-iodobenzene
SMILES C1=CC(=C(C=C1F)Br)I

Specifications

Density 1.33
Boiling Point 224°C to 225°C
Flash Point >110°C (230°F)
Refractive Index 1.629
Quantity 1 g
Beilstein 6288420
Sensitivity Light sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 300.9
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-4-fluoro-1-iodobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.