accessibility menu, dialog, popup

UserName

CH3H3COOOO

CAS RN 2033-24-1

IUPAC Name: 2,2-dimethyl-1,3-dioxane-4,6-dione
Synonyms: meldrum's acid
Molecular Weight (g/mol): 144.13
Molecular Formula: C6H8O4
InChi Key: GXHFUVWIGNLZSC-UHFFFAOYSA-N
SMILES: CC1(C)OC(=O)CC(=O)O1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (4)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)