missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%

Product Code. 10617352 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10617352 25 g Glass bottle 25g
10421821 100 g Glass bottle 100g
10647164 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10617352 Supplier Thermo Scientific Acros Supplier No. 198450250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2033-24-1 | C6H8O4 | 144.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2033-24-1
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.13
MDL Number MFCD00006638
InChI Key GXHFUVWIGNLZSC-UHFFFAOYSA-N
Synonym meldrum's acid, isopropylidene malonate, meldrums acid, 1,3-dioxane-4,6-dione, 2,2-dimethyl, sub-isopropyl malonate, 2,2-dimethyl-1,3-dioxan-4,6-dione, cycl-isopropylidene malonate, cyclic isopropylidene malonate, malonic acid cyclic isopropylidene ester, meldrum acid
PubChem CID 16249
IUPAC Name 2,2-dimethyl-1,3-dioxane-4,6-dione
SMILES CC1(OC(=O)CC(=O)O1)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 92°C to 96°C
Color Beige-Brown to Brown-White
Flash Point >365°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Fieser 08,313; 10,252; 11,311; 13,172
Merck Index 15, 5885
Solubility Information Solubility in water: 2.5g/100 mL (20°C). Other solubilities: slightly soluble in ethanol and chloroform
Formula Weight 144.13
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,2-Dimethyl-1,3-dioxane-4,6-dione
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2,2-Dimethyl-1,3-dioxane-4,6-dione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.