missing translation for 'onlineSavingsMsg'
Learn More

Isopropylidene malonate, 97%

Product Code. 11497467
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497467 500 g 500g
11477467 25 g 25g
11487467 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497467 Supplier Thermo Scientific Alfa Aesar Supplier No. A15603.36
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2033-24-1 | C6H8O4 | 144.126 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Isopropylidene malonate is an important reagent in synthetic chemistry and involved in the formation of new C-C bonds, rings, amides, esters, and acids. The presence of carbonyl groups at C-4 and C-6 positions favors the alkylation and acylation reactions due to its acidic nature. It is also employed as an antimicrobial agent. It is used in the preparation of 4-pyridyl-substituted heterocycles, 2-substituted indoles, isofraxidin and macrocyclic beta-keto lactones. Further, it is involved in the Knoevenagel condensation reaction with aldehydes.

Solubility
Soluble in water. Slightly soluble in ethanol and chloroform.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2033-24-1
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.126
MDL Number MFCD00006638
InChI Key GXHFUVWIGNLZSC-UHFFFAOYSA-N
Synonym meldrum's acid, isopropylidene malonate, meldrums acid, 1,3-dioxane-4,6-dione, 2,2-dimethyl, sub-isopropyl malonate, 2,2-dimethyl-1,3-dioxan-4,6-dione, cycl-isopropylidene malonate, cyclic isopropylidene malonate, malonic acid cyclic isopropylidene ester, meldrum acid
PubChem CID 16249
IUPAC Name 2,2-dimethyl-1,3-dioxane-4,6-dione
SMILES CC1(OC(=O)CC(=O)O1)C

Specifications

Melting Point 92°C to 96°C
Quantity 500 g
Beilstein 117310
Sensitivity Moisture sensitive
Merck Index 14,5817
Solubility Information Soluble in water. Slightly soluble in ethanol and chloroform.
Formula Weight 144.13
Percent Purity 98%
Chemical Name or Material Isopropylidene malonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.