accessibility menu, dialog, popup

UserName

BrOBr

CAS RN 2050-47-7

IUPAC Name: 1-bromo-4-(4-bromophenoxy)benzene
Synonyms: benzene, 1,1'-oxybis 4-bromo-
Molecular Weight (g/mol): 328
Molecular Formula: C12H8Br2O
InChi Key: YAWIAFUBXXPJMQ-UHFFFAOYSA-N
SMILES: BrC1=CC=C(OC2=CC=C(Br)C=C2)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results