accessibility menu, dialog, popup

UserName

H3COOClHNCH3O

CAS RN 20676-54-4

IUPAC Name: methyl 5-chloro-2-acetamidobenzoate
Synonyms: methyl 5-chloro-2-acetamidobenzoate
Molecular Weight (g/mol): 227.64
Molecular Formula: C10H10ClNO3
InChi Key: TVAAIYFBEWHVCV-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Cl)=CC=C1NC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results