missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-acetamido-5-chlorobenzoate, 97%, Thermo Scientific™

Product Code. 11422858
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11422858 250 g 250g
11412858 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11422858 Supplier Thermo Scientific Alfa Aesar Supplier No. A16826.30
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20676-54-4
Molecular Formula C10H10ClNO3
Molecular Weight (g/mol) 227.64
MDL Number MFCD00144759
InChI Key TVAAIYFBEWHVCV-UHFFFAOYSA-N
Synonym 2-acetamido-5-chlorobenzoic acid methyl ester, methyl 5-chloro-2-acetamidobenzoate, methyl n-acetyl-5-chloroanthranilate, n-acetyl-5-chloroanthranilic acid methyl ester, 2-acetylamino-5-chloro-benzoic acid methyl ester, methyl 2-acetylamino-5-chlorobenzoate, pubchem3337, acmc-1cioq, methyl2-acetamido-5-chlorobenzoate, methyl 2-acetamido-5-chloranyl-benzoate
PubChem CID 4640571
IUPAC Name methyl 2-acetamido-5-chlorobenzoate
SMILES COC(=O)C1=CC(Cl)=CC=C1NC(C)=O

Specifications

Melting Point 128°C to 131°C
Quantity 250 g
Beilstein 2808490
Formula Weight 227.65
Percent Purity 97%
Chemical Name or Material Methyl 2-acetamido-5-chlorobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.