accessibility menu, dialog, popup

UserName

CAS RN 20776-51-6

NH2BrOO

CAS RN 20776-51-6

IUPAC Name: 2-amino-3-bromobenzoate
Synonyms: 2-amino-3-bromobenzoate
Molecular Weight (g/mol): 215.03
Molecular Formula: C7H5BrNO2
InChi Key: SRIZNTFPBWRGPB-UHFFFAOYSA-M
SMILES: NC1=C(Br)C=CC=C1C([O-])=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results