missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-bromobenzoic acid, 97%

Product Code. 15428518
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15428518 5 g 5g
15418518 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15428518 Supplier Thermo Scientific Alfa Aesar Supplier No. H61925.06
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 20776-51-6 | C7H5BrNO2 | 215.03 g/mol

Used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediates.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20776-51-6
Molecular Formula C7H5BrNO2
Molecular Weight (g/mol) 215.03
MDL Number MFCD03618453
InChI Key SRIZNTFPBWRGPB-UHFFFAOYSA-M
Synonym 3-bromoanthralic acid, 2-amino-3-bromo-benzoic acid, benzoic acid, 2-amino-3-bromo, 3-bromoanthranilic acid, 2-amino-3-bromobenzoicacid, 2-amino-3-bromo benzoic acid, buttpark 49\07-48, 2-amino-3-bromo-benzoicacid, zlchem 426
PubChem CID 270259
IUPAC Name 2-amino-3-bromobenzoic acid
SMILES NC1=C(Br)C=CC=C1C([O-])=O

Specifications

Melting Point 174°C to 178°C
Color White to Yellow
Flash Point 161°C (322°F)
Quantity 5 g
Beilstein 2936928
Formula Weight 216.03
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Amino-3-bromobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.