CAS RN 2097971-11-2
CAS RN 2097971-11-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Tocris Bioscience™ VH 032 amide-PEG3-amine
PROTACs (Proteolysis Targeting Chimeras)
2
MedChemExpress (S,R,S)-AHPC-PEG3-NH2 hydrochloride
MedChemExpress (S,R,S)-AHPC-PEG3-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and 3-unit PEG linker used in PROTAC technology.
3