CAS RN 2098836-45-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ AT 1
(+)-JQ1 based PROTAC with selectivity for BRD4
2
BRD4 degrader AT1, MedChemExpress
MedChemExpress BRD4 degrader AT1 is a PROTAC connected by ligands for von Hippel-Lindau and BRD4 as a highly selective Brd4 degrader, with a Kd of 44 nM for Brd4BD2 in cells.