accessibility menu, dialog, popup

UserName

CAS RN 210110-90-0

H3CNOHHOOHOHHCl

CAS RN 210110-90-0

IUPAC Name: 1-butyl-2-(hydroxymethyl)piperidine-3,4,5-triol hydrochloride
Synonyms: 1-butyl-2-(hydroxymethyl)piperidine-3,4,5-triol hydrochloride
Molecular Weight (g/mol): 255.74
Molecular Formula: C10H22ClNO4
InChi Key: QPAFAUYWVZMWPR-UHFFFAOYNA-N
SMILES: Cl.CCCCN1CC(O)C(O)C(O)C1CO
Color 
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results