accessibility menu, dialog, popup

UserName

CAS RN 210353-53-0

IUPAC Name: 7-[(4Z)-3-(aminomethyl)-4-(methoxyimino)pyrrolidin-1-yl]-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid; methanesulfonic acid
Synonyms: gemifloxacin mesylate
Molecular Weight (g/mol): 485.49
Molecular Formula: C19H24FN5O7S
InChi Key: JIYMVSQRGZEYAX-CWUUNJJBNA-N
SMILES: CS(O)(=O)=O.CO\N=C1/CN(CC1CN)C1=C(F)C=C2C(=O)C(=CN(C3CC3)C2=N1)C(O)=O
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (3)
Color 
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results