accessibility menu, dialog, popup

UserName

H3COOHOOCH3

CAS RN 2107-70-2

IUPAC Name: 3-(3,4-dimethoxyphenyl)propanoic acid
Synonyms: 3-(3,4-dimethoxyphenyl)propanoic acid
Molecular Weight (g/mol): 210.23
Molecular Formula: C11H14O4
InChi Key: LHHKQWQTBCTDQM-UHFFFAOYSA-N
SMILES: COC1=CC=C(CCC(O)=O)C=C1OC
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)