missing translation for 'onlineSavingsMsg'
Learn More

3-(3,4-Dimethoxyphenyl)propionic acid, 98%

Product Code. 11446877
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11446877 25 g 25g
11436877 5 g 5g
11456877 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11446877 Supplier Thermo Scientific Alfa Aesar Supplier No. A14325.14
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2107-70-2 | C11H14O4 | 210.229 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2107-70-2
Molecular Formula C11H14O4
Molecular Weight (g/mol) 210.229
MDL Number MFCD00002774
InChI Key LHHKQWQTBCTDQM-UHFFFAOYSA-N
Synonym 3-3,4-dimethoxyphenyl propionic acid, 3-3,4-dimethoxyphenyl propanoic acid, 3,4-dimethoxyhydrocinnamic acid, benzenepropanoic acid, 3,4-dimethoxy, 3-3,4-dimethoxyphenyl-propionic acid, 3-3,4-dimethoxy-phenyl-propionic acid, unii-6xo32zsp1d, 6xo32zsp1d, 3-3',4'-dimethoxyphenyl propanoic acid, 3-3,4-dimethoxyphenyl propionicacid
PubChem CID 75019
ChEBI CHEBI:44235
IUPAC Name 3-(3,4-dimethoxyphenyl)propanoic acid
SMILES COC1=C(C=C(C=C1)CCC(=O)O)OC

Specifications

Melting Point 96°C to 99°C
Quantity 25 g
Beilstein 2696272
Formula Weight 210.23
Percent Purity 98%
Chemical Name or Material 3-(3,4-Dimethoxyphenyl)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.