CAS RN 21098-11-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
8-(4-Chlorobutyl)-8-azaspiro[4.5]-decane-7,9-dione, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
8-(4-Chlorobutyl)-8-azaspiro[4.5]decane-7,9-dione, TRC
CAS: 21098-11-3 Molecular Formula: C13 H20 Cl N O2 Molecular Weight (g/mol): 257.76 Synonym: 8-(4-Chlorobutyl)-8-azaspiro[4.5]-decane-7,9-dione,Buspirone Hydrochloride Imp. L (EP) IUPAC Name: 8-(4-chlorobutyl)-8-azaspiro[4.5]decane-7,9-dione SMILES: ClCCCCN1C(=O)CC2(CCCC2)CC1=O